Learn More
Sigma Aldrich Fine Chemicals Biosciences NorellR Sample Vault Serie
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z69318996EA
Encompass
for Bruker(R) SampleJet(TM) robotic high-throughput automated systems. NMR tubes with NorLoc(TM) caps are designed to be used with 96 position carriers using 103.5 mm (4 in.) long NMR tubes with open port caps or 178 mm (7 in.) long NMR tubes with closed port caps. NorLoc caps have superior holding and sealing capabilities which eliminate cap/tube failure in your instrument.Patent pending design incorporates a blue band positioned on the NMR tube that aligns with the base of the cap indicating a properly locked position. The NorLoc cap and Sample Vault Series tube is an integrated closure system for fail-safe delivery of your sample to the magnet. Tubes are available in both 3 mm and 5 mm diameters for use up to 700 MHz and up to 950 MHz.Engineered for new generation high-throughput lab automation systemsTwo precision types for up to 700 MHz and up to 950 MHz spectrometersSuperior NorLoc cap attaches semi-permanently for multiple use and for critical applicationsC
Catalog No. 50-290-5092