Learn More
Medchemexpress LLC HY-13715 500mg Medchemexpress, Norepinephrine CAS:51-41-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13715500MG
Encompass_Preferred
Medchemexpress, HY-13715 500mg Norepinephrine CAS:51-41-2 Norepinephrine (Levarterenol) is a β 1 -selective adrenergic receptor agonist with EC 50 of 5.37 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2679