Learn More
Medchemexpress LLC Norepinephrine | 51-41-2 | MFCD00025592 | 98.1% | C8H11NO3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13715R10MG
Encompass_Preferred
Norepinephrine analytical standard (CAS 51-41-2) is a high-purity reference material for analytical and research use. It is intended for calibration, method development, and quantitative analysis in chromatographic and mass spectrometric assays. Molecular formula C8H11NO3; example batch purity 98.08%.
  • High purity suitable for analytical calibration and quantitation.
  • Compatible with HPLC and LC-MS workflows.
  • Available in small vial sizes for standard preparation.
  • Offered in free base and common salt forms for method flexibility.
  • Provided with batch-specific certificate of analysis for traceability.
Catalog No. 50-002-59014