Learn More
Cayman Chemical NorepInphrInbItartrat 250mg
SDP
Supplier:  Cayman Chemical 16673250 MG
Encompass_Preferred
A natural neurotransmitter and hormone that acts as an agonist of adrenergic receptors (Ki values are 330, 56, and 740 nM for α1, α2, and β1 adrenoceptors, respectively)
Catalog No. 50-296-6297