Learn More
Cayman Chemical -NorepInphrInhydrochlo 100mg
SDP
Supplier:  Cayman Chemical 35580100 MG
Encompass_Preferred
A racemic mixture of of (–)-norepinephrine and (+)-norepinephrine; induces cAMP accumulation in rat cerebral cortical membranes at 1-100 µM; induces contraction of bovine anterior cerebral, middle cerebral, and internal carotid arterial strips (EC50s = 0.91, 0.92, and 0.87 µM, respectively) but induces relaxation of bovine posterior cerebral arterial strips (EC50 = 0.95 µM)
Catalog No. 50-291-3225