Learn More
Cayman Chemical NorethIndron 1g
SDP
Supplier:  Cayman Chemical 209411 G
Encompass_Preferred
A synthetic progestin; inhibits the estrous cycle in female mice at 0.5 mg/animal per day; induces tumor progression and metastasis in a BT474 breast cancer mouse xenograft model
Catalog No. 50-298-5478