Learn More
Medchemexpress LLC Norethynodrel | 68-23-5 | 90.53% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB13411MG
Encompass_Preferred
Norethynodrel is a 19-nortestosterone derivative used as a progestagen for researching inhibition of ovulation. It is also a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Derivative of 19-nortestosterone.
  • Used as a progestagen.
  • Can be used for researching inhibition of ovulation.
  • Click chemistry reagent.
  • Contains an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
Catalog No. 50-004-12012