Learn More
Medchemexpress LLC (+)-norfenfluramine hydrochloride | 37936-89-3 | MFCD00871809 | 99.8% | 239.67 g/mol | C10H13ClF3N | 10 MG
SDP
Supplier:  Medchemexpress LLC HY118010A10MG
Encompass_Preferred
(+)-Norfenfluramine hydrochloride is the chiral hydrochloride salt of norfenfluramine, a hepatic metabolite of fenfluramine, provided as a high-purity research reagent for receptor pharmacology and analytical applications.
  • Provides high-purity reference material for receptor and metabolic studies.
  • Acts as a selective 5-HT2B receptor agonist for pharmacological assays.
  • Supplied as a small research-scale quantity suitable for analytical and preparative use.
  • Solid white to off-white form that is stable under recommended storage conditions.
Catalog No. 50-003-07683