Learn More
Medchemexpress LLC Norfloxacin | 70458-96-7 | 98.31% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB013210G
Encompass_Preferred
Norfloxacin (MK-0366) is a broad-spectrum antibiotic effective against both Gram-positive and Gram-negative bacteria. It operates by inhibiting DNA gyrase and topoisomerase IV, enzymes crucial for bacterial DNA separation, thus hindering cell division. This synthetic chemotherapeutic antibacterial agent is occasionally used for treating common and complicated urinary tract infections.
  • Broad-spectrum antibiotic
  • Inhibits DNA gyrase and topoisomerase IV
  • Active against Gram-positive and Gram-negative bacteria
  • Used to treat urinary tract infections
Catalog No. 50-004-50868