Learn More
Medchemexpress LLC Norfloxacin | 70458-96-7 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0132R100MG
Encompass_Preferred
Norfloxacin (Standard) is an analytical standard intended for research and analytical applications. This broad-spectrum antibiotic is active against both Gram-positive and Gram-negative bacteria, functioning by inhibiting DNA gyrase. It is commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Active against Gram-positive and Gram-negative bacteria
  • Inhibits DNA gyrase
  • Suitable for HPLC, GC, and MS research experiments
Catalog No. 50-004-50875