Learn More
Medchemexpress LLC Norfloxacin-d5 | 1015856-57-1 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0132S1MG
Encompass_Preferred
Norfloxacin-d5 is a deuterium labeled Norfloxacin, functioning as a broad-spectrum antibiotic active against both Gram-positive and Gram-negative bacteria by inhibiting DNA gyrase. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-50923