Learn More
Medchemexpress LLC Norfloxacin (MK-0366) | 70458-96-7 | MFCD00079532 | 98.3% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB01325G
Encompass_Preferred
Norfloxacin (MK-0366) is a broad-spectrum antibiotic effective against both Gram-positive and Gram-negative bacteria. Its mechanism of action involves inhibiting DNA gyrase, a type II topoisomerase, and topoisomerase IV. These enzymes are crucial for the separation of bacterial DNA, and their inhibition by Norfloxacin consequently prevents bacterial cell division. This synthetic chemotherapeutic antibacterial agent is occasionally utilized to treat both common and complicated urinary tract infections. The product is intended for research use only.
  • Broad-spectrum antibiotic
  • Effective against Gram-positive and Gram-negative bacteria
  • Inhibits DNA gyrase and topoisomerase IV
  • Prevents bacterial cell division
  • Suitable for research applications
Catalog No. 50-004-50506