Learn More
Medchemexpress LLC Norfluoxetine-d5 hydrochloride | 1188265-34-0 | 99.3% | 336.79 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW011235S1MG
Encompass_Preferred
Norfluoxetine-d5 (hydrochloride) is a deuterium-labeled compound. It can be used as a tracer and as an internal standard for quantitative analysis. Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-52846