Learn More
Cayman Chemical NorfluoxetInhydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 159005 MG
Encompass_Preferred
An active metabolite of fluoxetine; inhibits 5-HT uptake in rat brain synaptosomal membrane preparations (Ki = 44.7 nM) and isolated human platelets (IC50 = ~15 nM); has been found in the tissues of fish exposed to wastewater effluent
Catalog No. 50-297-2245