Learn More
American Radiolabeled Chemicals Inc NORGESTREL 3H 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 198050UC
Encompass_Preferred
Norgestrel, D-15,16-3H 50uCi in ethanol, 1 mCi/ml, 50-70 Ci/mmol, M.W. 312.45, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-964