Learn More
Cayman Chemical NorhyodeoxycholIc AcId 10mg
SDP
Supplier:  Cayman Chemical 900344010 MG
Encompass_Preferred
A synthetic bile acid and a derivative of HDCA; an intermediate in the synthesis of 3β-sulfooxy-7β-hydroxy-24-nor-5-cholen-23-oic acid
Catalog No. 50-289-5782