Learn More
Discovery Life Sciences Normal BMMC
SDP
Supplier:  Discovery Life Sciences 100300
Encompass
BMMCs Bone Marrow Mononuclear Cells are isolated from bone marrow aspirates and supplied as viably cryopreserved immune and progenitor cells commonly used in hematologic cancer research immune profiling and cell therapy development
Catalog No. 50-008-04122