Learn More
Cayman Chemical -NornIcotIn-d4 5mg
SDP
Supplier:  Cayman Chemical 352425 MG
Encompass_Preferred
An internal standard for the quantification of (±)-nornicotine by GC- or LC-MS
Catalog No. 50-291-3452