Learn More
Medchemexpress LLC (±)-Nornicotine | 5746-86-1 | 148.20 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00211210MM/1ML
Encompass_Preferred
(±)-Nornicotine is a major metabolite of Nicotine. It is a partial nAChRs agonist, specifically activating receptor subtypes containing α7 and α6 subunits. It can be used in the research of atherosclerosis, Alzheimer's disease, and schizophrenia.
  • Promotes cell migration, disrupts adherens junctions and tight junctions
  • Induces F-actin depolymerization
  • Maintains self-administration behavior in rats
Catalog No. 50-004-47824