Learn More
Medchemexpress LLC (±)-Nornicotine | 5746-86-1 | 148.20 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW002112100MG
Encompass_Preferred
(±)-Nornicotine is a major metabolite of Nicotine. It functions as a partial nAChRs agonist, specifically activating receptor subtypes containing α7 and α6 subunits. This compound disrupts β-catenin and ZO-1, and induces F-actin depolymerization. It supports self-administration behavior and is utilized in research for conditions such as atherosclerosis, Alzheimer's disease, and schizophrenia.
  • Promotes cell migration in human endothelial cells
  • Disrupts adherens junctions (β-catenin) and tight junctions (ZO-1)
  • Induces F-actin depolymerization
  • Maintains self-administration behavior in Sprague-Dawley rats
Catalog No. 50-004-47344