Learn More
Medchemexpress LLC (±)-Nornicotine | 5746-86-1 | 148.20 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW00211250MG
Encompass_Preferred
(±)-Nornicotine is a major metabolite of Nicotine that acts as a partial nAChRs agonist, specifically activating receptor subtypes containing α7 and α6 subunits. This compound disrupts β-catenin and ZO-1, and induces F-actin depolymerization. It supports self-administration behavior and can be utilized in research concerning atherosclerosis, Alzheimer's disease, and schizophrenia.
  • Acts as a partial nAChRs agonist
  • Activates receptor subtypes containing α7 and α6 subunits
  • Disrupts β-catenin and ZO-1
  • Induces F-actin depolymerization
  • Supports self-administration behavior
  • Suitable for research concerning atherosclerosis, Alzheimer's disease, and schizophrenia
  • Promotes cell migration and disrupts adherens/tight junctions in vitro
  • Maintains self-administration behavior in vivo
Catalog No. 50-004-47374