Learn More
Medchemexpress LLC (±)-Nornicotine-d4 | 66148-18-3 | 152.23 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW002112S1MG
Encompass_Preferred
(±)-Nornicotine-d4 is the deuterium labeled version of (±)-Nornicotine, a major metabolite of Nicotine. Deuterium labeling incorporates stable heavy isotopes into drug molecules, which can be used as tracers for quantitation during drug development and may affect pharmacokinetic and metabolic profiles. This product is suitable for research in areas such as atherosclerosis, Alzheimer's disease, and schizophrenia.
  • Deuterium labeled compound
  • Major metabolite of Nicotine
  • Partial nAChRs agonist
  • Activates receptor subtypes containing α7 and α6 subunits
  • Disrupts β-catenin and ZO-1
  • Induces F-actin depolymerization
  • Supports self-administration behavior
  • Stable under recommended storage conditions
Catalog No. 50-004-47104