Learn More
Medchemexpress LLC (±)-Nornicotine-d4 | 66148-18-3 | 152.23 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW002112S100MG
Encompass_Preferred
(±)-Nornicotine-d4 is the deuterium labeled (±)-Nornicotine, a major metabolite of Nicotine. It functions as a partial nAChRs agonist, specifically activating receptor subtypes containing α7 and α6 subunits. This compound is suitable for research applications in various fields.
  • Disrupts β-catenin and ZO-1
  • Induces F-actin depolymerization
  • Supports self-administration behavior
  • Applicable in research for atherosclerosis, Alzheimer's disease, and schizophrenia
Catalog No. 50-004-47522