Learn More
Alpco Ltd Norovirus RT-PCR Kit
SDP
Supplier:  Alpco Ltd 86NVIHU100
Encompass
The Norovirus RT-PCR Kit is a multiplex real-time RT-PCR assay for the direct, qualitative detection of norovirus RNA (genogroup GI and GII) in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.
Catalog No. NC2284608