Learn More
U.S. Pharmacopeia Norphenylephrine hydrochloride, 4779-94-6, MFCD00012879, 25 mg
Supplier:  U.S. Pharmacopeia 1473206
Encompass
Linear Formula: HOC6H4CH(CH2NH2)OH · HCl, Molecular Weight: 189.64, mp: 162-164 °C, Synonym: α-Aminomethyl-3-hydroxybenzyl alcohol hydrochloride, 3,α-Dihydroxyphenethylamine hydrochloride, Norfenefrine hydrochloride.
Catalog No. NC1590279