Learn More
Saint Gobain Performance Plastics NORPRENE CHEMICAL 50 FT. 1/PK
Supplier:  Saint Gobain Performance Plastics AD300007
Encompass_Preferred
Saint-Gobain Norprene Chemical Tubing Diameter Inner 0.13 in. Diameter Outer 1/4 in. Maximum Pressure 33 psi Thickness 0.0625 in.
Catalog No. 50-206-8984