Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P High-Performance Precision Pump Tubing, Norprene Food, I/P 89; 25 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0640289
Encompass
Masterflex Norprene Food High-Performance Precision pump tubing is formulated to withstand extreme temperatures and will not deteriorate in demanding food and beverage applications. This tubing is compatible with virtually all commercial clears and sanitizers and can be sterilized via autoclave without impacted overall service life. Ideal for food and beverage applications, this tubing complies with FDA, NSF, and 3-A standards.
Catalog No. NC0095953