Learn More
Cayman Chemical Norpslocn 5mg
SDP
Supplier:  Cayman Chemical 235865 MG
Encompass_Preferred
An analytical reference standard categorized as a tryptamine; has been found in P. cubensis; a potential metabolite of baeocystin based on the published metabolism of psilocybin; intended for research and forensic applications
Catalog No. 50-299-5279