Learn More
Cayman Chemical NorsufentnIl 10mg
SDP
Supplier:  Cayman Chemical 900114210 MG
Encompass_Preferred
A metabolite of sufentanil, produced by oxidative N-dealkylation in the liver by CYP450 isoforms; intended for forensic applications
Catalog No. 50-289-4107