Learn More
TARGETMOL CHEMICALS INC Nortopixantrone HCl 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T281891MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Nortopixantrone HCl (BBR 3438) is a novel 9-azacyclonopyrazole that shows antitumor activity in a human prostate cancer model. Purity 98.03%
Catalog No. 50-287-9428