Learn More
Medchemexpress LLC Nortriptyline (hydrochloride) (Standard) | 894-71-3 | 99.98% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB1417
Encompass_Preferred
Nortriptyline hydrochloride, also known as Desmethylamitriptyline hydrochloride, is a tricyclic antidepressant and the primary active metabolite of Amitriptyline. It functions as a potent autophagy inhibitor and demonstrates anticancer properties. This product is intended for research and analytical applications.
  • Analytical standard of Nortriptyline (hydrochloride).
  • Intended for research and analytical applications.
  • Main active metabolite of Amitriptyline.
  • Tricyclic antidepressant.
  • Potent autophagy inhibitor.
  • Exhibits anticancer effects.
  • Reduces viability of TCCSUP and mouse MBT-2 bladder cancer cells.
  • Induces cell cycle arrest and apoptosis in TCCSUP and MBT-2 cells.
  • Inhibits the growth of bladder tumors in mice.
Catalog No. 50-004-14124