Learn More
Cayman Chemical NortrIptylInhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1590410 MG
Encompass_Preferred
A tricyclic antidepressant that blocks norepinephrine and serotonin transporters (KDs = 4.4 and 18 nM, respectively) more potently than the dopamine transporter (KD = 1.1 µM)
Catalog No. 50-296-9565