Learn More
Medchemexpress LLC Norvancomycin monohydrochloride | 213997-73-0 | 95.3% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1924
Encompass_Preferred
Norvancomycin hydrochloride is a cell wall synthesis inhibitor that targets peptidoglycan precursors of Gram-positive bacteria and does not cross the blood-brain barrier. It competitively binds to peptidoglycan precursors, irreversibly inhibiting bacterial cell wall synthesis and exhibiting antibacterial activity. It is primarily used in the study of Gram-positive bacterial infections, particularly those caused by methicillin-resistant *Staphylococcus aureus* (MRSA) and methicillin-resistant *Staphylococcus epidermidis* (MRSE).
  • Inhibits cell wall synthesis
  • Targets peptidoglycan precursors of gram-positive bacteria
  • Does not cross the blood-brain barrier
  • Used in the study of gram-positive bacterial infections
  • Can be integrated into bionic calcium phosphate coatings on titanium implants
Catalog No. 50-004-12037