Learn More
Medchemexpress LLC Norverapamil hydrochloride | 67812-42-4 | MFCD00078605 | 99.8% | 477.04 g·mol⁻¹ | C26H37ClN2O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10075050MG
Encompass_Preferred
Norverapamil hydrochloride is the N-demethylated metabolite of verapamil that functions as an L-type calcium channel blocker and a P-glycoprotein (P-gp) function inhibitor. It is supplied as a solid and as a DMSO solution for use as a reference standard and reagent in pharmacology, transporter assays, and mechanistic studies.
  • N-demethylated metabolite of verapamil.
  • Acts as an L-type calcium channel blocker and P-gp inhibitor.
  • Available as solid and 10 mM DMSO solution.
  • High purity suitable for research applications.
  • Used in pharmacological and transporter functional studies.
Catalog No. 50-002-12188