Learn More
Medchemexpress LLC Norverapamil hydrochloride ((±)-norverapamil hydrochloride) | 67812-42-4 | 99.8% | 477.04 g/mol | C26H37ClN2O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10075025MG
Encompass_Preferred
Norverapamil hydrochloride is the N-demethylated metabolite of verapamil that functions as an L-type calcium channel blocker and a P-glycoprotein (P-gp) function inhibitor. It is provided as a high-purity research reagent for pharmacology, transporter studies, and drug-metabolism investigations.
  • N-demethylated metabolite of verapamil.
  • L-type calcium channel blocker activity.
  • P-glycoprotein (P-gp) function inhibitor.
  • High purity suitable for research applications.
  • Soluble in water and DMSO, facilitating assay formulation.
  • Recommended sealed storage at 4°C; solution stability at -80°C and -20°C.
Catalog No. 50-002-12186