Learn More
TARGETMOL CHEMICALS INC NOS-IN-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T217005MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NOS-IN-1 (2-Imino-4-methylpiperidine acetate) is a potent and orally active inhibitor of NO synthase (NOS) isoforms. NOS-IN-1 exhibits IC50s of 0.1 uM 0.2 uM and 1.1uM for human iNOS (hiNOS) hnNOS and heNOS respectively. purity: 100%
Catalog No. 50-257-7206