Learn More
Santa Cruz Biotechnology NOS2 Antibody (C-11) | 200 µg/ml
Supplier:  Santa Cruz Biotechnology SC7271
Encompass
Anti-NOS2 Antibody (C-11) is a mouse monoclonal IgG1 κ NOS2 antibody, cited in 383 publications, provided at 200 µg/ml. Specific for an epitope mapping between amino acids 1126-1144 near the C-terminus of NOS2 of mouse origin. Anti-NOS2 Antibody (C-11) is recommended for detection of NOS2 (iNOS) of mouse, rat and human origin by WB, IP, IF, IHC(P), FCM and ELISA.
Catalog No. NC1827286