Learn More
LICOR Nose Cone Plugs for in vivo Imaging (pack of 6 small)
Supplier:  LICOR 9300800
Encompass_Preferred
Nose cone plugs are used to block anesthesia ports that are not being used for in vivo imaging. They are available in packs of six in either small plugs designed for mouse nose cones or large plugs designed for rat nose cones.
Catalog No. NC2225411