Learn More
Medchemexpress LLC Nosiheptide | 56377-79-8 | 97.1% | 1222.36 g/mol | C51H43N13O12S6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY107486100MG
Encompass_Preferred
Nosiheptide is a thiopeptide antibiotic produced by Streptomyces actuosus that inhibits bacterial protein synthesis by interfering with elongation factors. It is supplied as a research-grade solid for use in microbiology and biochemical studies probing translation and antibiotic mechanisms.
  • Thiopeptide antibiotic that inhibits bacterial protein synthesis.
  • Reported high purity suitable for research applications.
  • Available in multiple solid sizes for experimental flexibility.
  • Molecular formula and weight available for analytical reference.
  • Useful for studies of translation inhibition and antibiotic mechanism of action.
Catalog No. 50-002-39867