Learn More
Medchemexpress LLC Not found in product page or linked SDS | 1349637-14-4 | 98.2% | 491.06 | C30H35ClN2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16090110MG
Encompass_Preferred
CP-289,503 is a small-molecule inhibitor of the complement C5a receptor supplied for research use. It exhibits inhibitory activity with an IC50 of approximately 1 μM and is provided as a high-purity powder suitable for biochemical and cellular studies.
  • Inhibits complement C5a receptor (IC50 ≈ 1 μM).
  • High purity: 98.21%.
  • Molecular weight 491.06; formula C30H35ClN2O2.
  • Available in multiple pack sizes, including 10 MG.
  • Powder storage: -20°C (up to 3 years); in solvent: -80°C (up to 6 months).
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-88834