Learn More
Selleck Chemical LLC NOTA
SDP
Supplier:  Selleck Chemical LLC S69235MG
Encompass_Preferred
NOTA i e 1 4 7-triazacyclononane-1 4 7-triacetic acid is a bifunctional chelating agent which acts as the framework to construct PET imaging tools It is also used for probe design and signal amplification via the multivalent effect and also has potential applications as diagnostic gallium radiopharmaceuticals
Catalog No. 50-000-02486