Learn More
Medchemexpress LLC Notoginsenoside Fc | 88122-52-5 | 98.9% | 1211.38 g·mol-1 | C58H98O26 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN253110MG
Encompass_Preferred
Notoginsenoside Fc is a protopanaxadiol (PPD)-type saponin isolated from the leaves of Panax notoginseng. It has been reported to inhibit platelet aggregation and to accelerate reendothelialization after vascular injury by promoting autophagy, making it useful for biochemical and pharmacological research into platelet function and vascular repair mechanisms.
  • Ppd-type saponin isolated from Panax notoginseng.
  • Demonstrated to inhibit platelet aggregation.
  • Promotes reendothelialization by enhancing autophagy.
  • High reported purity (approximately 98.9%), suitable for research use.
  • Solid powder, off-white to light yellow; store sealed away from moisture and light.
Catalog No. 50-002-88934