Learn More
Medchemexpress LLC Notoginsenoside Fe | 88105-29-7 | MFCD30478378 | 99.9% | 917.13 | C47H80O17 | 10 MG

Supplier: Medchemexpress LLC HYN004610MG
Notoginsenoside Fe is a triterpene saponin isolated from Panax notoginseng, provided as a high-purity solid for research use. It is intended for biochemical and pharmacological studies, and supporting documentation for quality and safety is available.
- High purity (99.9%).
- Molecular weight 917.13 g/mol.
- Molecular formula C47H80O17.
- Supplied as a solid for convenient handling.
- Suitable for biochemical and pharmacological research applications.
- Certificate of analysis and safety data sheet available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.