Learn More
Medchemexpress LLC Notoginsenoside FP2 | 1004988-75-3 | 1 MG

Supplier: Medchemexpress LLC HYN43051MG
Notoginsenoside FP2 is a dammarane-Type Bisdesmoside isolated from the Fruit Pedicels of Panax notoginseng, and it has the potential to treat cardiovascular disease.
- Purity is 99.61%.
- Molecular weight is 1211.38.
- Molecular formula is C58H98O26.
- Appearance is a white to off-white solid.
- Classified as Terpenoids, Triterpenes.
- Derived from plants, specifically Araliaceae, Panax notoginseng (Burkill) F. H. Chen ex C. H. Chow.
- Soluble in DMSO at 100 mg/mL (82.55 mM), requiring ultrasonic treatment.
- Store at 4°C, protected from light. In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.