Learn More
Medchemexpress LLC Notoginsenoside FP2 | 1004988-75-3 | MFCD32197358 | 99.6% | 1211.38 g·mol-1 | C58H98O26 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN430510MG
Encompass_Preferred
Notoginsenoside FP2 is a natural dammarane-type bisdesmoside isolated from the fruit pedicels of Panax notoginseng and supplied for research use in pharmacological and biochemical studies related to cardiovascular activity.
  • Natural product isolated from Panax notoginseng.
  • High purity: 99.61% (manufacturer specification).
  • Well-characterized molecular formula C58H98O26 and molecular weight 1211.38 g·mol-1.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic treatment.
  • Available in small-scale research quantities suitable for experimental assays.
Catalog No. 50-002-89918