Learn More
Medchemexpress LLC Notoginsenoside R2 | 80418-25-3 | 770.99 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN09091MG
Encompass_Preferred
Notoginsenoside R2 is a newly isolated notoginsenoside from Panax notoginseng, showed neuroprotective effects against 6-OHDA-induced oxidative stress and apoptosis. For research use only. We do not sell to patients.
  • Neuroprotective effects against 6-OHDA-induced oxidative stress and apoptosis.
Catalog No. 50-004-15577