Learn More
Medchemexpress LLC Notoginsenoside R2 | 80418-25-3 | 770.99 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN09095MG
Encompass_Preferred
Notoginsenoside R2 is a newly isolated notoginsenoside from Panax notoginseng, which showed neuroprotective effects against 6-OHDA-induced oxidative stress and apoptosis.
  • For research use only.
  • We do not sell to patients.
Catalog No. 50-004-16485