Learn More
United States Biological Corporation Nourseothricin, (clonNAT), 1 Gram
Supplier:  United States Biological Corporation N5375741G
Encompass_Preferred
Notch 1 is a transmembrane protein functioning in the development and determination of cell-fate (1). During maturation, the Notch molecule is cleaved by furin-like convertase at the extracellular domain (2). Upon binding to a ligand such as Delta1, or upon extracellular calcium depletion, the C-terminal Notch 1 fragment dissociates from its N-terminal counterpart and is further cleaved between Gly1743 and Val1744 (3,4). The resulting activated cytosolic fragment translocates to the nucleus and activates Notch-related transcription. Applications: Suitable for use in Western Blot, Immunoprecipitation, and ELISA. Other applications not tested. Recommended Dilution: Western Blot: 1:1000 Incubate membrane with diluted antibody in 5% BSA, 1X TBS, 0.1% Tween-20 at 4C with gentle shaking, overnight. Immunoprecipitation: 1:50 Optimal dilutions to be determined by the researcher. Storage and Stability: May be stored at 4C for short-term only.
Catalog No. NC0861695