Learn More
AG Scientific Inc Nourseothricin Sulfate 1 G
Supplier:  AG Scientific Inc N26751GM
Encompass_Preferred
Dihydrogen sulfate form of the weekly basic antibiotic Nourseothricin consisting of components F, E and D. Effective against gram negative and gram positive bacteria. Also effective against mycobacteria, mycoplasms, protozoa, yeasts, viruses and plants.Nourseothricin sulfate is commonly used as a selection agent for recombinant yeast strains.
Catalog No. 50-226-8339