Learn More
Abcam Nourseothricin sulfate, 50MG
Supplier:  Abcam AB28540950MG
Encompass_Preferred
MW 600.6 Da, Purity >85%. Nourseothricin is group of a broad-spectrum antibiotic produced by Streptomyces variants; inhibits protein synthesis by inducing miscoding. Streptothricin antibiotic Nourseothricin (NTC) is used for a broad spectrum of viruses, bacteria and other unicellular or complex organisms.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-4878